missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates

Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model

Brand:  EMD Millipore™ MANMN6010

Code : 73

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11724694

  • £369.55 / Pack of 10


Estimated Shipment
09-02-2018
Add to basket

Description

Description

96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.

  • Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
  • Optically clear for use with an imaging microscope or other microscopic analysis
  • Can be coated with a thin layer of agar if necessary
  • Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer

Specifications

Specifications

MultiScreen Mesh
0.28cm2
Styrene
96
Non-sterile
Clear
Whole organism screening
Nylon Mesh
60μm
0.05 to 0.25mL
Videos