Learn More
Merck MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates
Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model
Brand:  Merck MANM10010
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11704694
  • £865.00 / Pack of 10
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.
  • Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
  • Optically clear for use with an imaging microscope or other microscopic analysis
  • Can be coated with a thin layer of agar if necessary
  • Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer
Specifications
Specifications
Standard
8 x 12
Clear
96
0.28cm2
10/Pk.
Nylon Mesh
100 μm
Styrene
Non-sterile
Whole organism screening