missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates
Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model
£687.00 - £865.00
Specifications
Format | Standard |
---|---|
Membrane | Nylon Mesh |
Array | 8 x 12 |
Color | Clear |
Material | Styrene |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11704694
|
Merck
MANM10010 |
100 μm |
£865.00
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10517285
|
Merck
MANMN2010 |
20 μm |
£780.00
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11714694
|
Merck
MANMN4010 |
40 μm |
£687.00
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11724694
|
Merck
MANMN6010 |
60 μm |
£829.00
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.
- Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
- Optically clear for use with an imaging microscope or other microscopic analysis
- Can be coated with a thin layer of agar if necessary
- Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer
Specifications
Standard | |
8 x 12 | |
Styrene | |
Non-sterile | |
10/Pk. |
Nylon Mesh | |
Clear | |
96 | |
Whole organism screening |