missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
Brand: Merck MSHAS4B10
Code : 73
Additional Details : Weight : 0.10000kg
Description
- Developed for high-throughput use with automated work stations
- Individually sealed wells prevent incubation crosstalk
- Removable underdrain allows access to the filters
- Rigid sidewalls improve handling
- Ample surfaces provided for bar code labels
- Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
Specifications
Standard | |
8 x 12 | |
Opaque | |
Barex | |
Sterile | |
Solid phase RIA, DNA hybridization, ELISA, protein binding |
Mixed Cellulose Esters | |
0.45 μm | |
MultiScreenHTS-HA Plate | |
96 | |
0.28 cm2 | |
10/Pk. |