Learn More
Merck MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
Brand: Merck MZFBN0W10
Code : 73
Additional Details : Weight : 0.10000kg
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSI™/SBS standards
Specifications
Standard | |
16 x 24 | |
Opaque, White Bottom | |
Barex | |
384 | |
0.09 cm2 | |
10/Pk. | |
1.28 cm | |
1.44 cm |
Glass Fiber, Polyester Mesh | |
1 μm | |
MultiScreenHTS-FB Plate | |
0.02 to 0.1 mL | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |