missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ MultiScreenHTS 384-Well Filter Plates

Use in automated, high throughput applications, including biological assays and sample preparation. EMD Millipore MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.

Brand:  EMD Millipore™ MZPHN0W50

Code : 73

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11714704

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.
  • Rigid sidewalls allows full compatibility with automated/robotic systems
  • Unique plate design and individually sealed filter discs prevent cross-talk between wells
  • Opaque plastic lids prevents light transfer between wells during direct scintillation counting
  • Barcoding available
  • Meet ANSI™/SBS standards
Specifications

Specifications

MultiScreenHTS-PH Plate
1.28 x 8.55 x 1.44cm
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Styrene
384
50
0.02 to 0.1mL
8.55cm
Opaque
0.09cm2
1.28cm
Negatively Charged Phosphocellulose, Polyester Mesh
0.65μm
Non-sterile
1.44cm
Videos
Documentation