missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ MultiScreen™ 96-Well Assay Plates for Sample Preparation

For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications

Brand:  EMD Millipore™ MADVN6550

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10370031

  • £899.81 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Formats are suitable for removal of large particulates, resin-based separations, solvent applications, and protein/DNA concentration or purification. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold

Choice of filter materials

  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes

Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.

Specifications

Specifications

MultiScreen-DV Plate
0.28cm2
Styrene
96
Non-sterile
Clear
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase assays, bead, resin based assays, sample prep
Hydrophilic PVDF
0.65μm
0.05 to 0.25mL
Videos
View accessory products