missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Montage™ Plasmid Miniprep96 Kits
Generate high-quality plasmid DNA in 50% less time than traditional methods
Brand: Merck LSKP09624
Code : 72
Additional Details : Weight : 0.80000kg
Includes: All necessary reagents and disposables
Requires : MultiScreen™ Vacuum Manifold with underdrain support grid (MAVM 096 0R)
Description
- Typical yield from 1mL of bacterial culture is 4 to 6μg
- 96-well kit uses unique separation technology Simple, 25-minute protocol yields clean and reproducible DNA for sensitive downstream applications Collection of purified sample from the top of the plate and lack of centrifugation steps allow compatibility with automated systems
Specifications
Microplate | |
All necessary reagents and disposables | |
Kit |
Plasmid Miniprep, BAC Prep | |
24 x 96 Preps |