Learn More
Merck Montage™ Plasmid Miniprep96 Kits
Generate high-quality plasmid DNA in 50% less time than traditional methods
Brand: Merck LSKP09604
Code : 72
Additional Details : Weight : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Includes: All necessary reagents and disposables
Requires : MultiScreen™ Vacuum Manifold with underdrain support grid (MAVM 096 0R)
Description
- Typical yield from 1mL of bacterial culture is 4 to 6μg
- 96-well kit uses unique separation technology Simple, 25-minute protocol yields clean and reproducible DNA for sensitive downstream applications Collection of purified sample from the top of the plate and lack of centrifugation steps allow compatibility with automated systems
Specifications
Microplate | |
All necessary reagents and disposables | |
Kit |
Plasmid Miniprep, BAC Prep | |
4 x 96 Preps |
Safety and Handling
- Plasmid MiniPrep96 Kit, 24 Plates
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P234-Keep only in original container.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.