missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Montage™ Antibody Purification Kits with Prosep™-A and Prosep-G Media

Provide a centrifugal method for fast and easy purification of antibodies from serum, ascites, or cell culture supernatants

Brand:  EMD Millipore™ LSK2ABA60

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10143693

  • £756.00 / Pack of 6


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

The Prosep family of media has a long history of success for large-scale bioprocessing applications including Prosep-A for industrial monoclonal antibody purification. Montage Kits deliver that same level of performance and reliability for research-scale antibody purification.

  • Purify up to 10mL of sample in less than 60 minutes Yield up to 20mg of purified antibody
  • Utilize Prosep high-capacity media, which offers rapid transport and a very high dynamic capacity for IgG

Specifications

Specifications

Red
No
1600μL
Porous Glass
Antibody purification
2° to 8°C
6 Pack
Videos
Documentation

Technical Documents