EMD Millipore™ Millipak™ Disposable Filter Units

Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Overview
Brand: EMD Millipore™

Manufacturer Part Number: MPGL04GH2

X2 Filter unit Millipak 40 disposable and sterilized by gamma radiation 6mm (1/4inch) stepped hose

UNSPSC: 41104914

Code: 73

Additional Details:
Additional Details: Weight: 3.40190kg



Product Code. 13118093

Quantity Price
1 £ 225.5 / Pack of 2
Estimated Shipment
15-02-2017
Add to basket


Description and Specification

Specification

Connectors 0.25 in./6.35mm stepped hose barb inlet and outlet with bell
Product Type Filter unit
Disposable Yes
Filtration Area 200cm2
Flow Rate 2.8L/min. at 1.75 bar
Inlet Pressure 75psi at 25°C
Diameter (Metric) Outer 7.6cm
Material (Membrane) PVDF
Sterile Yes
Pore Size (Metric) 0.22μm
Volume (Metric) 20L
Material Polycarbonate
Filter Type Durapore™
For Use With (Application) Removal of particles and microorganisms from liquids and gases
Length (Metric) 8.9cm
Quantity (No. per Case) 2 Pack

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity