missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millipak™ Disposable Filter Units

Removes particles and microorganisms from liquids and gases with pump-driven filter. Merck Millipore™ Millipak™ Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.

Brand:  EMD Millipore™ MPGL06GF2

Code : 73

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 12871626

  • £250.00 / Pack of 2


Estimated Shipment
13-02-2018
Add to basket

Description

Description

Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.

  • Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
  • Point-of-use filter for high-purity water
  • Ideally suited for high value-added applications
  • Durapore™ PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
  • Presterilized units are gamma irradiated
  • Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
  • 100% integrity tested to guarantee integrity

Specifications

Specifications

7.6cm
0.22μm
Polycarbonate
Filter unit
Durapore™
7.6L/min. at 2 bar
75psi at 25°C
PVDF
Yes
2 Pack
0.75 in./19.05mm sanitary flange inlet and outlet
Yes
300cm2
Removal of particles and microorganisms from liquids and gases
9.4cm
30L
Videos