Learn More
EMD Millipore™ Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC
Non sterile Universal bottletop vacuum filter
Brand:  EMD Millipore™ SJFHM4710
Code : 71
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10772161
  • £95.50 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
The Millicup vacuum-driven bottletop filter unit is a disposable device that provides a fast, effective, and safe means to filter mobile phases and running buffers used in instrumentation analyses (for example, HPLC). The Millicup vacuum-driven bottletop filter unit eliminates the risk of contaminant carry-over from solvent to solvent. Unlike reusable filter holders that must be washed and assembled before each use, Millicup units require no assembly or clean-up. If accidentally dropped, the Millicup vacuum-driven bottletop filter unit will not break, ensuring a safe environment in which to filter solvents and other hazardous solutions.
Specifications
Specifications
HPLC
Hydrophobic PTFE
Millicup™ Filter Unit
12cm
0.45μm
Videos