missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC

Non sterile Universal bottletop vacuum filter

Brand:  EMD Millipore™ SJLHM4710

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10278282

  • £91.00 / Pack of 10


Estimated Shipment
14-02-2018
Add to basket

Description

Description

The Millicup vacuum-driven bottletop filter unit is a disposable device that provides a fast, effective, and safe means to filter mobile phases and running buffers used in instrumentation analyses (for example, HPLC). The Millicup vacuum-driven bottletop filter unit eliminates the risk of contaminant carry-over from solvent to solvent. Unlike reusable filter holders that must be washed and assembled before each use, Millicup units require no assembly or clean-up. If accidentally dropped, the Millicup vacuum-driven bottletop filter unit will not break, ensuring a safe environment in which to filter solvents and other hazardous solutions.
Specifications

Specifications

HPLC
Hydrophilic PTFE
Millicup™ Filter Unit
12cm
0.45μm
Videos