Learn More
EMD Millipore™ Milli-Q™ Ultrapure Water Systems Accessory, Cartridges
Choose from a wide selection of replacement cartridges
Brand:  EMD Millipore™ QTUM000VX
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12999423
  • £685.33 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Filter Types
  • Activated carbon
  • Ion exchange
  • Membrane
Specifications
Specifications
Quantum VX
Polishing Cartridge
Milli-Q Systems
Activated Carbon, Ion Exchange
Videos