Learn More
EMD Millipore™ Milli-Q™ Ultrapure Water Systems Accessory, Cartridges
Choose from a wide selection of replacement cartridges
Brand:  EMD Millipore™ QTUM000IX
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12989423
  • £484.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Filter Types
  • Activated carbon
  • Ion exchange
  • Membrane
Specifications
Specifications
Quantum IX
Polishing Cartridge
Milli-Q Systems
Ion exchange
Videos