Learn More
EMD Millipore™ Milli-Q™ Advantage A10 Water Purification Packs
For use with Advantage A10 system
Brand:  EMD Millipore™ QGARDT2X1
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12909353
  • £440.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Specifications
Specifications
Deionized water
Milli-Q™ Advantage 10 water purification system
Ion exchange, activated Carbon, depth filtration
Q-Gard T2 purification pack
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Videos