missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Sterile Syringe Filters: Durapore™ PVFD Membrane

Designed for sterilizing and clarifying sterile solutions. EMD Millipore Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.1, 0.22, 0.45 and 5.0µm pore sizes; medical-grade filters available.

Brand:  EMD Millipore™ SLHV033RK

Code : 71

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10038031

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • 100% integrity tested, individually packaged, and sterilized
  • Female Luer-Lok™ inlet and male Luer slip outlet
  • Manufactured in a controlled environment using an automated process
  • Yellow color-coded band clearly indicates which membrane is inside
  • ≤0.22µm filters are sterilizing grade
  • Larger pore sizes are for clarification and prefiltration
  • Include a Certificate of Quality

Durapore™ (PVDF) membrane

  • Lowest protein binding sterilization/clarification of biological solutions
  • Made of polyvinylidene fluoride (PVFD)
  • Gamma irradiation sterilized
Compliance
  • Meet USP Class VI Biological tests for plastics
  • All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Specifications

Specifications

Syringe Filter
Durapore Hydrophilic Polyvinylidene Fluoride (PVDF)
Use for sterilization/clarification of water, buffers, and salt solutions
<200mL
Female Luer Lok
45°C
0.45μm
Modified Acrylic
33mm
145psi
<100μL
Meets USP Class VI Biological tests for plastics. All 33mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S.
Male Luer Slip
1,000/pk.
Yes
Yellow
Videos
Documentation