Learn More
EMD Millipore™ Millex™ Nonsterile Syringe Filters with PVC Housing
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Brand:  EMD Millipore™ SLHA02510
Code : 09
Additional Details : Weight : 3.86000kg
 View more versions of this product
Product Code. 10627131
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • With female Luer-Lok™ inlet and male Luer slip outlet
  • Available in a variety of membrane types
MCE Membrane
  • Mixed cellulose ester membrane for sterilization/clarification of water, buffers, and salt solutions
PVDF Membrane
  • Lowest protein binding
  • Use for aqueous and mild organic solutions
Mitex™-SV
  • Compatible with more aggressive organic solutions and high temperature conditions
Specifications
Specifications
Syringe Filter
MCE
PVC
No
25mm
For sterilization or clarification of water, buffers, and salt solutions
0.45μm
1000/Pk.
Videos