missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - PVDF Membrane - 4mm Diameter

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  EMD Millipore™ SLGVR04NK

Code : 71

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10645961

  • £1255.00 / Pack of 1000


Estimated Shipment
08-02-2018
Add to basket

Description

Description

  • With female Luer-Lok™ inlet and male Luer slip outlet
  • Lowest protein binding
  • Use for aqueous and mild organic solutions

Specifications

Specifications

Syringe Filter
PVDF Membrane
HDPE
1000/Pk.
4.0mm
Use for aqueous and mild organic solutions
0.22μm
Videos
Documentation