missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™ Syringe Filter, Fluoropore™ PTFE, Hydrophobic, 0.2 μm

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  Merck SLFG025NS

Product Code. 10024470

  • £291.00 / Pack of 50
Estimated Shipment: 14-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Overview of Non-Sterile Millex™ Syringe Filters with Durapore™ (PVDF) Membrane
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Compatible with aqueous and mild organic solutions
  • Housing materials are either polypropylene or high density polyethylene (HDPE), offering low levels of extractables and broad chemical compatibility

Applications:
 Sample Filtration Prior to UHPLC, HPLC, LC-MS, and Mass Spec; Solvent Filtration; Filtration of Biological Samples and Protein Solutions

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
0.2 μm
Analytical sample prep
Female Luer-Lok™
50Pack
100 mL
25 mm
PTFE
Non-sterile
HDPE
Male Luer slip
Syringe Filter
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers