missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  EMD Millipore™ SLCR025NB

Code : 71

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10379970

  • £780.00 / Pack of 250


Estimated Shipment
16-02-2018
Add to basket

Description

Description

  • Broad chemical compatibility Choose for filtering aqueous and organic solutions Low protein binding

Specifications

Specifications

Syringe Filter
Polytetrafluoroethylene (PTFE)
Polyethylene
25/Pk.
25mm
For filtering aqueous and organic solutions
0.45μm
Videos
Documentation