missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  EMD Millipore™ SLCR025NK

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10136760

  • £1600.00 / Pack of 1000


Estimated Shipment
11-05-2018
Add to basket

Description

Description

  • Broad chemical compatibility Choose for filtering aqueous and organic solutions Low protein binding

Specifications

Specifications

Syringe Filter
Hydrophilic LCR (PTFE)
Polyethylene
1000/Pk.
25mm
For filtering aqueous and organic solutions
0.45μm
Videos
Documentation