missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  Merck Millipore SLCR025NS

Code : 71

Additional Details : Weight : 0.16544kg

 View more versions of this product

Product Code. 10044350

  • £250.00 / Pack of 50


In Stock 7
Add to basket

Description

Description

  • Broad chemical compatibility Choose for filtering aqueous and organic solutions Low protein binding

Specifications

Specifications

Syringe Filter
Polytetrafluoroethylene (PTFE)
Polyethylene
50/Pk.
25mm
For filtering aqueous and organic solutions
0.45μm
Videos
Documents