missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophilic PTFE

For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines

Brand:  EMD Millipore™ SLLGR04NL

Code : 71

Additional Details : Weight : 0.13160kg

 View more versions of this product

Product Code. 10350261

  • £168.00 / Pack of 100


In Stock 8
Add to basket

Description

Description

  • Broad chemical compatibility Choose for filtering aqueous and organic solutions Low protein binding

Specifications

Specifications

Syringe Filter
Polytetrafluoroethylene (PTFE)
Polyethylene
100/Pk.
4.0mm
For filtering aqueous and organic solutions
0.2μm
Videos
Documentation