missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Millex™ Filters: Inlet and Outlet for Silicone Tubing

Protect vacuum sources from contamination and moisture damage

Brand:  EMD Millipore™ SLFG85010

Code : 71

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10238090

  • £115.00 / Pack of 10


In Stock 13
Add to basket

Description

Description

Stepped hose barb with female Luer slip interior.

  • With hydrophobic PTFE membranes
  • Polypropylene housing (except for SLFG-025-LS and -50, which have PVC housing)
  • Variety of inlet/outlet combinations, including stepped hose barb and 1/8 NPT
  • Nonsterile, autoclavable, bidirectional

Applications:

  • Protect vacuum sources from contamination and moisture damage
  • Sterilize ambient air entering fermenters
  • Filter incubator gas to protect against contamination
  • Filter nitrogen and air used to dry liquid samples

Specifications

Specifications

50mm
PTFE
No
10/Pk.
0.2μm
Polypropylene
Yes
Protect vacuum sources from contamination and moisture damage; Sterilize ambient air entering fermenters; Filter incubator gas to protect against contamination; Filter nitrogen and air used to dry liquid samples
Videos
Documentation