Learn More
EMD Millipore™ Microfil™ S Filtration Devices
Fast, dependable alternative to conventional multi-step methods
Brand:  EMD Millipore™ MVHAWGS24
Code : 72
Additional Details : Weight : 1.20000kg
 View more versions of this product
Product Code. 13167853
  • £185.33 / Pack of 24
Estimated Shipment
14-06-2018
Add to basket
Description
Description
Devices consist of a funnel, protective lid, and filtration support so routine microbiological testing is fast and easy.
  • Ensure consistent recovery of microorganisms
  • Smooth, hydrophobic funnel surface repels aqueous sample residues to ensure that microorganisms are collected on the membrane, not on funnel walls
  • Convex support shape eliminates pleating and distortion of membrane due to expansion
  • Three membrane selections offer versatility for your specific application
Benefits
  • Presterilized, individually packaged No membrane handling prior to filtering
  • No filter holders, funnels and attachments to assemble and sterilize
  • No sterilizing the filter support after each filtration Protective lid reduces external contamination risk Pre-assembled to speed sampling and shorten preparation Easy-to-remove funnel and membrane reduce test time
Specifications
Specifications
MCE
0.45μm
EMD Millipore Microfil S Filtration Devices
Videos
Documentation
Technical Documents