missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Microfil™ Funnels

Available in different colors

Brand:  EMD Millipore™ MZHABG251

Code : 72

Additional Details : Weight : 2.70000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 12363318

  • £329.00 / Pack of 150


In Stock 8
Add to basket


Description

Description

  • 0.22μm pore size for specific test regulations
  • 0.45μm for general microbiological analysis
  • 0.7μm for chlorinated effluents
  • 0.8μm for yeast or molds applications

Specifications

Specifications

Black
0.45μm
250mL
Microfil Filtration System Funnels
EZ-Pak membranes
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Kit: 150xMicrofil 250ml funnel + 150x0.45

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

Supplemental information
  • MIXTURE LIST-Contains¬†: nitrocellulose