missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Microfil™ Funnels

Available in different colors

Brand:  EMD Millipore™ MZHAWG101

Code : 72

Additional Details : Weight : 1.61680kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

 View more versions of this product

Product Code. 10518452

  • £268.00 / Pack of 150


In Stock 9
Add to basket

Description

Description

  • 0.22μm pore size for specific test regulations
  • 0.45μm for general microbiological analysis
  • 0.7μm for chlorinated effluents
  • 0.8μm for yeast or molds applications

Specifications

Specifications

White
Gridded
EZ-Pak membranes
Microfil Filtration System Funnels
0.45μm
100mL
Videos
Documentation