Learn More
Merck Microcon-30 kDa Centrifugal Filter Unit with Ultracel-30 Membrane
This 0.5 mL centrifugal filter enables the concentration of biomolecules with a membrane NMWL of 30 kDa. Micron centrifugal filters are ideal for DNA & protein concentration & desalting.
Brand: Merck MRCF0R030
471.41 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.28080kg
Description
ideal for DNA concentration and desalting applications.
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel™ membrane, Microcon™ filters offer:•Typical recoveries of greater than 95%, even for dilute solutions •Reverse spin to maximize recovery, even in the smallest samples •Convenient storage of filtrate or concentrated sample in standard microfuge tube •Concentration factors up to 100X Features:•High recovery for small volumes with reverse spin (concentration factor less than 20X) •Low-binding Ultracel™ membrane •Fast processing Sample applications:•Recovery of genomic DNA for forensic applications •Concentration and desalting of nucleic acids (high recovery alternative to ethanol precipitation) •Removal of primers from amplified DNA •Easy filtrate recovery, exploiting the flat membrane designSpecifications
Clear | |
Regenerated Cellulose | |
Non-sterile | |
Regenerated Cellulose (RC) | |
10 mL | |
4.5 cm |
Single-use | |
30 kDa | |
0.32 cm2 | |
Centrifugal Filters | |
1.8 in. | |
12.3 mm |