Learn More
Merck MF-Millipore™ Membrane Filter, 1.2 μm, Gridded
Mixed cellulose esters (MCE) membrane, hydrophilic
Brand: Merck RAWG02500
Code : 71
Additional Details : Weight : 0.10000kg
Description
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
Not Autoclavable | |
MCE | |
Non-sterile | |
For biological and environmental monitoring applications | |
Gridded | |
100/Pk. | |
Hydrophilic | |
25 mm |
White | |
1.2 μm | |
270 mL/min x cm2 | |
Hydrophilic | |
100Pack | |
Filter Membranes | |
25 mm |
Safety and Handling
- MCE 1.2UM WH GD 25MM 100/PK
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
- MIXTURE LIST-Contains : octylphenyl polyethoxyethanol