Learn More
EMD Millipore™ M Air T™ Agar Cassettes
Ready-to-use cassettes, prefilled with gamma-sterilized medium
Brand:  EMD Millipore™ ATSMTTD60
Code : 73
Additional Details : Weight : 3.10000kg
 View more versions of this product
Product Code. 12841606
  • £248.00 / Pack of 60
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Fast and reliable results with maximum recovery
  • Large agar amount and unique micro-perforated sieve allow air sampling of up to 1 cubic meter
  • Unit-to-unit consistency of agar surface shape
  • Double-sleeve packaged
  • Reproducible results
Compliance
USP
Specifications
Specifications
M air T Air Testing System
Trypticase Soy Agar media
USP
M air T Cassettes
For the total count of airborne microorganisms in pharmaceutical clean rooms, hospitals, food and beverage applications
Videos
Documentation
Technical Documents