Learn More
Merck™ Individual Cartridges for Milli-Q™ and Milli-RO™ Bowl-Style Systems
For Milli-Q
Brand:  Merck™ CPMB01202
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10060222
  • £463.00 / Pack of 2
Estimated Shipment
10-01-2020
Description
Description
For Milli-Q Bowl systems
Specifications
Specifications
Ion-Ex Cartridge
Replacement Cartridge
Depth filtration
12 in. (30cm) cartridge removes ionized impurities from water pretreated by reverse osmosis, deionization or distillation. Constructed of pure polypropylene with thermally bonded endcaps to reduce the risk of extractables contamination.
Milli-Q™ and Milli-RO™ bowl-style systems