missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Immobilon™-PSQ PVDF Membranes
For Western blots and protein sequencing
Brand: Merck ISEQ20200
Code : 72
Additional Details : Weight : 0.10000kg
Description
Immobilon-PSQ Membrane:Specifications
20 cm | |
For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins | |
Immobilon-PSQ | |
20 x 20 cm |
20 cm | |
Sheet | |
0.2 μm |