missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™-PSQ PVDF Membranes

For Western blots and protein sequencing

Brand:  Merck ISEQ09120

140.39 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10593984

  • £159.00 / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • Specifications

    Specifications

    9 cm
    For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    Immobilon-PSQ
    9 x 12 cm
    12 cm
    Sheet
    0.2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers