missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Hydrophobic 6mm Edge Membranes

For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations

Brand:  EMD Millipore™ HAEP0470W

Code : 71

Additional Details : Weight : 0.02000kg Transport : UN number : 3270 Chem class : 1 Pack group : II

 View more versions of this product

Product Code. 12861566

  • £174.67 / Pack of 100


Estimated Shipment
02-03-2018
Add to basket

Specifications

Specifications

White
0.45μm
Plain
Microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations, filling machine operations, sterility testing
Mixed Cellulose Ester
No
4.7cm
Hydrophobic Edge Membrane
Videos