missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for YT30090HW ZHE Hazardous Waste System
Accessories for filter holders for sample preparation.
Brand: Merck XX3002567
Code : 72
Additional Details : Weight : 0.10000kg
Description
Accessories for filter holders for sample preparation.Specifications
NPTF to M | |
Laboratory Filter Holders or Cyclones | |
0.125 in. |
Water Purification System | |
Nickel/Chrome plated | |
0.32 cm |