Learn More
EMD Millipore Fitting Adapter
Brand:  EMD Millipore XX3002561
UNSPSC : 41115700
Code : 72
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10578591
  • £62.46 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
NPTM to M
Water Purification System
0.32cm
Luer Lock Adapter
Nickel/Chrome plated