missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Fast-Trap™ Adenovirus and Lentivirus Purification and Concentration Kits

Quick, easy membrane-based method for laboratory-scale virus purification

Brand:  EMD Millipore™ FTAV00003

Code : NEW

Additional Details : Weight : 0.99000kg

 View more versions of this product

Product Code. 10156263

  • £582.67 / 1 set


Estimated Shipment
02-03-2018
Add to basket

Alert:

These kits permit quick purification of infectious agents which should be handled under BSL2 safety precautions. Wear hand, eye, face, and body personal protective equipment (PPE) when using this kit. All virus purification steps (except water bath incubation and centrifugation) should be conducted in a biosafety cabinet.

Disclaimers:

For laboratory use only; not for diagnostic or in vivo use.


Specifications

Specifications

1 x 1013 virus particles
Adenovirus purification and concentration
<2 hr.
3.0
Fast trap adenovirus purification and concentration kit
Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
Room Temperature
Videos