missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Fast-Trap™ Adenovirus and Lentivirus Purification and Concentration Kits

Quick, easy membrane-based method for laboratory-scale virus purification

Brand:  EMD Millipore™ FTLV00003

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11720343

  • £673.33 / 1 set


Call for latest availability
+44 (0)1509 555 500
Add to basket

Alert:

These kits permit quick purification of infectious agents which should be handled under BSL2 safety precautions. Wear hand, eye, face, and body personal protective equipment (PPE) when using this kit. All virus purification steps (except water bath incubation and centrifugation) should be conducted in a biosafety cabinet.


Description

Description

Fast-Trap Purification

  • Saves time—protocol can be completed in less than two hours
  • High yield—recover up to 70% of virus particles
  • Dependable—simple protocol ensures quality results
  • Easy—Steriflip™ filters eliminate messy purification steps and expensive equipment
  • Safe—use a closed vacuum device to eliminate potential spills

Specifications

Specifications

2 x 108 virus particles
Adenovirus purification and concentration
<2 hr.
3.0
Fast trap adenovirus purification and concentration kit
Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
Room Temperature
Videos

For laboratory use only; not for diagnostic or in vivo use.