missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ EZ-Pak™ Membranes

Used for general microbiological analysis. Millipore™ EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.

Brand:  EMD Millipore™ ezgswg474

Code : 72

Additional Details : Weight : 0.12000kg Transport : UN number : 3270 Chem class : 1 Pack group : II

 View more versions of this product

Product Code. 10577034

  • £370.00 / Pack of 600


Estimated Shipment
05-06-2018
Add to basket

Description

Description

Press lever to uncover half of membrane surface for easy retrieval with forceps; other half remains in protective packaging. Unused membranes remain safely packed in pleated band.

  • Mixed Cellulose Ester
  • Sterile and grid surface
  • Used for general microbiological analysis
  • Certified to meet standard methods for the examination of Water and wastewater requirements

Specifications

Specifications

Certified to meet standard methods for the examination of Water and wastewater requirements
Forceps
0.22μm
Grid
Particle counting, specific test regulations
White
Mixed Cellulose Ester
Yes
4.7cm
EZ-Pak Membrane
Videos