missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore EZ-Pak™ Membranes

Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.

Brand:  Merck Millipore EZHAWG474

Code : 72

Additional Details : Weight : 0.52110kg

 View more versions of this product

Product Code. 10059880

  • £493.00 / Pack of 600


In stock, ready to ship: 3
Add to basket

Description

Description

Used for general microbiological analysis. EZ-Pak™ Membranes are sterile, ready-to-use membranes that eliminate contamination risk and save time.

Press lever to uncover half of membrane surface for easy retrieval with forceps; other half remains in protective packaging. Unused membranes remain safely packed in pleated band.

  • Mixed Cellulose Ester
  • Sterile and grid surface
  • Used for general microbiological analysis
  • Certified to meet standard methods for the examination of Water and wastewater requirements

Specifications

Specifications

Certified to meet standard methods for the examination of Water and wastewater requirements
1.85 in.
General microbiological analysis and particle counting
Grid
Mixed Cellulose Ester
Sterile
White
4.7 cm
Forceps
EZ-Pak Membrane
0.45 μm
Videos
SDS
Documents