missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Express™ PLUS Membrane Filters

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

Brand:  EMD Millipore™ GPWP01300

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10400041

  • £74.00 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Hydrophilic, high-throughput, low-protein-binding
  • Thickness: ≥150μm and ≤200μm
  • Sterilization: by autoclave (121°C at 1 bar), EO or gamma
  • Bacterial endotoxins: 0.5 EU/mL
  • Gravimetric extractables: <1.5%
  • Flowrate: 40mL/min. cm2 at 2 bar, 25°C

Applications:

Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.

Specifications

Specifications

0.2μm
13mm
Yes
Hydrophilic
Polyethersulfone (PES)
White
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Videos