missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ EcoStand™ Tube Holder

Features a user-friendly design for both 15 and 50mL (0.51 and 1.69 oz.) tube formats

Brand:  EMD Millipore™ SC50TB001

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11740433

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

No
White
4.0
4 × 50mL tubes
Tube Holder
ASTM D6400, EN 13432
4 x 50mL tubes
Room temperature
Corn-based bioplastic resin
No
Videos