Learn More
Merck Centriprep™ Centrifugal Filter Units
For processing high-solute samples
Brand: Merck 4307
Code : 72
Additional Details : Weight : 0.10000kg
Description
For processing high-solute samples
Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.Features & Benefits:
Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Specifications
Clear | |
Ultracel-YM Regenerated Cellulose | |
Non-sterile | |
Regenerated Cellulose (RC) | |
Centrifugal Filters | |
135 mm |
Single-use | |
30 kDa | |
2.84 cm2 | |
96Pack | |
12 to 15 mL |