missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Centriprep™ Centrifugal Filter Units

For processing high-solute samples

Brand:  EMD Millipore™ 4305

Code : 72

Additional Details : Weight : 4.76270kg

 View more versions of this product

Product Code. 10402521

  • £808.00 / Pack of 96


Estimated Shipment
23-02-2018
Add to basket

Description

Description

  • Fast sample processing
  • High-recovery, Ultracel™-YM membrane
  • Ideal for concentrating and purifying particle-laden solutions or high concentrations
  • Separate low MW solutes from fermentation broths, cell culture media, and cell lysates
  • Unique inverse flow mode with large deadstop
  • Use in 50mL tube swinging-bucket or fixed-angle rotor

Specifications

Specifications

Ultracel-YM Regenerated Cellulose
10,000 d
2.84cm2
135mm
96/Pk.
12 to 15mL
Videos