EMD Millipore Amicon™ Ultra-2.0 Centrifugal Filter Units

Reverse spin capability provides consistent recoveries

Overview
Brand: EMD Millipore

Manufacturer Part Number: UFC210024PL

UNSPSC: 41120000

Code: X2

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11532144

Quantity Price
1 £ 107.0 / Pack of 24
Estimated Shipment
27-04-2017
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

MWCO 100,000 d
Surface Area 1cm2
Length (Metric) 4.99cm
For Use With (Application) Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange
Max. Force 14,000xG for concentration spin; 1000xG for reverse spin
Quantity 24/Pk.
Volume (Metric) 2.0mL
Membrane Material Regenerated Cellulose
Includes Two microcentrifuge tubes

  • Concentrate 2mL down to 15 – 70μL
  • True engineered dead stop provides consistent recovery without spinning to dryness
  • Reverse spin aids in sample recovery, especially with small, dilute samples
  • Vertical membrane design aligns with the filtrate for less clogging and faster filtering
  • Ultracel low-binding membrane combines with the innovative vertical housing to provide fast sample processing without sacrificing recovery
  • Fast processing, with 10- to 30-minute spin times

Applications:

  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
  • Desalting and buffer exchange

Alternative Products