missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Pro Purification System with Ultra-0.5 Device, 3 Kda

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

Brand:  Merck ACS500324

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15207956

  • £348.00 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
Specifications

Specifications

Single-use
Non-sterile
24 Amicon Ultra NMWCO 3 kDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Centrifugal Filters
12 cm
3 KDa
Regenerated Cellulose (RC)
24Pack
15 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers