missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Amicon™ Bioseparations Stirred Cells†

Concentrate, desalt, and purify aqueous biological solutions

Brand:  EMD Millipore™ 5121

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10402511

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Requires : Magnetic stirrer required for operation.

Description

Description

Process from 3mL up to 400mL of biological solutions using either concentration or diafiltration operating modes.

  • Sturdy construction; autoclavable Consist of clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings, and black aluminum stand
Concentration Mode
  • Gas pressure is applied directly to ultrafiltration cell Solutes above membrane molecular weight cutoff are retained in cell, while water and solutes below cutoff pass into filtrate
Diafiltration Mode
    Ultrafiltration cell is connected to auxiliary reservoir containing diafiltrate solution; reservoir is then gas pressurized Fluid volume and macrosolute concentration remain constant as filtrate is replaced by diafiltrate solution Provides a simple means for rapid microsolute exchange
  • Typically used as a substitute for dialysis

Applications:

Perform concentration, desalting, recovery or exclusion; Recover monoclonal antibodies and growth factors from tissue culture supernatants; Concentration and buffer exchange of chromatography fractions

Specifications

Specifications

Yes
0.03 to 0.3 oz., 1 to 10mL
2.5cm
4.1cm2
7.7cm
Nylon cap and tube fitting assembly, Polysulfone cylinder and membrane holder, Acetal and Polysulfone stirring assembly, Silicone Rubber O-rings, Polyethelene pressure tube, Tygon tubing filtrate tube, Anodized Aluminum or Nylon retaining stand
200μL
Clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings and black aluminum stand, connections for 0.25 in. dia. pressure tubing
1 to 10mL
0.05 to 10g%
2.38 x 2.38 x 3 in. (6 x 6 x 7.7cm)
Protein purification, protein concentration, protein desalting
6cm
8010
6cm
Videos
Documentation

Technical Documents

View accessory products