missing translation for 'onlineSavingsMsg'
Learn More

Merck Filter Aervent-50 50mm both sides with hose for 6.5 to 9.5mm

Brand:  Merck MTGR85000

Filter Aervent-50 50mm both sides with hose for 6.5 to 9.5mm

Code : 73

Additional Details : Weight : 2.75000kg

Product Code. 12861646

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.